Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164680.1 | NP_001158152.1 | 3 | 1121 | ||
NM_001837.3 | NP_001828.1 | 3 | 1140 | |||
NM_178328.1 | NP_847898.1 | 3 | 1130 | |||
NM_178329.2 | NP_847899.1 | 2 | 1061 | |||
XM_006712960.3 | XP_006713023.1 | 3 | 1167 | |||
XM_011533335.2 | XP_011531637.1 | 3 | 1247 | |||
XM_017005685.1 | XP_016861174.1 | 3 | 2050 | |||
XM_017005686.1 | XP_016861175.1 | 4 | 2231 | |||
GenBank mRNA | AF026535.1 | - | 1 | 892 | ||
BC110297.1 | - | 2 | 970 | 5212541 | ||
BC130318.1 | - | 2 | 972 | 40146349 | ||
BC130320.1 | - | 2 | 972 | 40146351 | ||
U28694.1 | - | 1 | 923 |