Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TDRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198795.1 | NP_942090.1 | 18 | 2567 | ||
XM_005269978.2 | XP_005270035.1 | 18 | 2548 | |||
XM_005269979.2 | XP_005270036.1 | 18 | 2581 | |||
XM_011539959.2 | XP_011538261.1 | 18 | 2920 | |||
XM_011539960.2 | XP_011538262.1 | 18 | 2510 | |||
XM_011539961.2 | XP_011538263.1 | 18 | 2492 | |||
XM_011539962.1 | XP_011538264.1 | 18 | 2874 | |||
XM_011539963.1 | XP_011538265.1 | 18 | 2581 | |||
XM_011539964.1 | XP_011538266.1 | 17 | 2410 | |||
XM_011539966.1 | XP_011538268.1 | 17 | 2410 | |||
XM_017016414.1 | XP_016871903.1 | 17 | 2437 | |||
XM_017016415.1 | XP_016871904.1 | 16 | 2266 | |||
GenBank mRNA | AF285606.1 | - | 11 | 1536 | ||
AK024735.1 | - | 8 | 1087 | |||
AK122916.1 | - | 18 | 2577 | |||
AK304331.1 | - | 9 | 1364 | |||
AL833340.1 | - | 18 | 2569 | |||
BC035010.2 | - | 18 | 2873 | |||
BX648664.1 | - | 18 | 2513 |