Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_054027.4 | NP_473368.1 | 11 | 1682 | ||
XM_017009644.1 | XP_016865133.1 | 11 | 1282 | |||
GenBank mRNA | AB046801.1 | - | 11 | 1512 | ||
AF274753.1 | - | 11 | 1419 | |||
AK001799.1 | - | 7 | 807 | |||
AK130676.1 | - | 6 | 646 | |||
AK222631.1 | - | 1380 | ||||
AK315012.1 | - | 11 | 1380 | |||
AY358503.1 | - | 11 | 1433 | |||
BC009835.2 | - | 11 | 1607 | 3927236 | ||
BC014526.2 | - | 11 | 1407 | 3837372 |