Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306188.1 | NP_001293117.1 | 2 | 405 | ||
NM_019556.2 | NP_062456.1 | 2 | 405 | |||
XM_011531366.2 | XP_011529668.1 | 2 | 359 | |||
XM_017029653.1 | XP_016885142.1 | 3 | 598 | |||
XM_017029654.1 | XP_016885143.1 | 2 | 359 | |||
GenBank mRNA | AF086333.1 | - | 2 | 299 | ||
AK299074.1 | - | 2 | 318 | |||
AK299172.1 | - | 1 | 195 | |||
AK307196.1 | - | 2 | 308 | |||
AK316570.1 | - | 2 | 280 | |||
AL137163.1 | - | 2 | 328 | |||
BC005700.1 | - | 2 | 308 | 3450155 | ||
CB997365.1 | - | 2 | 313 | |||
CN428237.1 | - | 2 | 407 | |||
DQ323992.1 | - | 1 | 141 |