Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MOSPD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306188.1 | NP_001293117.1 | 3 | 433 | ||
NM_019556.2 | NP_062456.1 | 3 | 433 | |||
XM_011531366.2 | XP_011529668.1 | 3 | 387 | |||
XM_017029653.1 | XP_016885142.1 | 4 | 626 | |||
XM_017029654.1 | XP_016885143.1 | 3 | 387 | |||
GenBank mRNA | AF086333.1 | - | 2 | 441 | ||
AK299074.1 | - | 3 | 346 | |||
AK299172.1 | - | 2 | 223 | |||
AK316570.1 | - | 3 | 308 | |||
AL137163.1 | - | 3 | 356 | |||
BC005700.1 | - | 3 | 336 | 3450155 | ||
CB997365.1 | - | 3 | 341 | |||
CN428237.1 | - | 3 | 435 | |||
DQ323992.1 | - | 2 | 169 |