Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM214A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286495.1 | NP_001273424.1 | 11 | 3216 | ||
NM_019600.3 | NP_062546.2 | 12 | 3215 | |||
NR_104457.1 | 12 | 3452 | ||||
XM_005254547.2 | XP_005254604.1 | 12 | 3178 | |||
XM_005254548.2 | XP_005254605.1 | 12 | 3697 | |||
XM_011521789.2 | XP_011520091.1 | 12 | 3244 | |||
XM_011521790.1 | XP_011520092.1 | 10 | 3147 | |||
GenBank mRNA | AB037791.1 | - | 12 | 3193 | ||
AK001842.1 | - | 7 | 1058 | |||
AK292917.1 | - | 12 | 3204 | |||
AK293920.1 | - | 11 | 3206 | |||
AK308049.1 | - | 11 | 3023 | |||
AL832519.1 | - | 1 | 553 | |||
BC040548.1 | - | 12 | 3452 | 5288206 | ||
BC058839.1 | - | 7 | 1011 | |||
BC073908.1 | - | 1 | 139 | |||
BC083498.1 | - | 5 | 548 | |||
BC109127.1 | - | 11 | 3146 | 40036501 | ||
BC109128.2 | - | 11 | 3146 | 40036502 |