Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRTAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304782.1 | NP_001291711.1 | 5 | 579 | ||
NM_019604.3 | NP_062550.2 | 10 | 1127 | |||
XM_011542900.2 | XP_011541202.1 | 9 | 1053 | |||
GenBank mRNA | AB209830.1 | - | 5 | 488 | ||
AF001622.1 | - | 10 | 1078 | |||
AK293470.1 | - | 5 | 508 | |||
AK313578.1 | - | 10 | 1125 | |||
BC070266.1 | - | 10 | 1127 | 30418882 | ||
BX118379.1 | - | 4 | 294 |