Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127244.1 | NP_001120716.1 | 3 | 950 | ||
NM_001127245.1 | NP_001120717.1 | 2 | 574 | |||
NM_019594.3 | NP_062540.2 | 3 | 681 | |||
XM_005252095.2 | XP_005252152.1 | 3 | 811 | |||
XM_005252096.3 | XP_005252153.1 | 3 | 1974 | |||
XM_006717186.2 | XP_006717249.1 | 4 | 1026 | |||
XM_011518863.1 | XP_011517165.1 | 3 | 617 | |||
XM_011518864.2 | XP_011517166.1 | 4 | 735 | |||
XM_011518865.2 | XP_011517167.1 | 2 | 453 | |||
GenBank mRNA | AB037858.1 | - | 2 | 490 | ||
AK024649.1 | - | 2 | 511 | |||
AK289842.1 | - | 3 | 950 | |||
AY143166.1 | - | 1 | 427 | |||
BC051322.1 | - | 3 | 593 | 6250713 | ||
BX376144.2 | - | 3 | 625 |