Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GKN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019617.3 | NP_062563.3 | 2, 3 | 168 | ||
GenBank mRNA | AJ966789.1 | - | 2, 3 | 168 | ||
AY139184.1 | - | 2, 3 | 106 | |||
AY359050.1 | - | 2, 3 | 92 | |||
BC059778.1 | - | 2, 3 | 94 |