Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL36G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278568.1 | NP_001265497.1 | 4 | 425 | ||
NM_019618.3 | NP_062564.1 | 5 | 530 | |||
GenBank mRNA | AF200492.1 | - | 5 | 525 | ||
AF206696.1 | - | 5 | 517 | |||
AY359111.1 | - | 441 | ||||
BC096721.1 | - | 4 | 475 | 40003618 | ||
BC098130.1 | - | 4 | 475 | 40003616 | ||
BC098155.1 | - | 4 | 475 | 40003617 | ||
BC098337.1 | - | 4 | 475 | 40003612 |