Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCR6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004367.5 | NP_004358.2 | 3 | 726 | ||
NM_031409.3 | NP_113597.2 | 3 | 989 | |||
GenBank mRNA | AK223494.1 | - | 3 | 645 | ||
AK293015.1 | - | 3 | 663 | |||
AY242126.1 | - | 1 | 421 | |||
BC037960.1 | - | 3 | 627 | 5748098 | ||
DN996219.1 | - | 3 | 714 | |||
U60000.1 | - | 1 | 421 | |||
U68030.1 | - | 4 | 685 |