Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRMT8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256536.1 | NP_001243465.1 | 9 | 1083 | ||
NM_019854.4 | NP_062828.3 | 9 | 1492 | |||
XM_017019644.1 | XP_016875133.1 | 9 | 1417 | |||
XM_017019645.1 | XP_016875134.1 | 7 | 1175 | |||
GenBank mRNA | AB209027.1 | - | 8 | 4991 | ||
AF263539.1 | - | 8 | 957 | |||
AK026786.1 | - | 11 | 1683 | |||
AK093651.1 | - | 5 | 1134 | |||
AK315619.1 | - | 9 | 1084 | |||
BC022458.1 | - | 9 | 1099 | 4796524 |