Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144002.1 | NP_001137474.1 | 2 | 817 | ||
NM_019857.4 | NP_062831.3 | 2 | 370 | |||
NM_175859.2 | NP_787055.1 | 2 | 456 | |||
XM_005274562.3 | XP_005274619.1 | 2 | 233 | |||
XM_006724503.3 | XP_006724566.1 | 3 | 408 | |||
XM_011545545.2 | XP_011543847.1 | 2 | 229 | |||
GenBank mRNA | AF226667.1 | - | 2 | 273 | ||
AK023549.1 | - | 2 | 328 | |||
AK024070.1 | - | 2 | 311 | |||
AK125332.1 | - | 2 | 817 | |||
AK125348.1 | - | 2 | 447 | |||
BC034986.1 | - | 2 | 435 | 5268973 | ||
DA741994.1 | - | 2 | 233 | |||
DA759070.1 | - | 2 | 371 |