Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTPS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144002.1 | NP_001137474.1 | 4 | 1148 | ||
NM_019857.4 | NP_062831.3 | 4 | 701 | |||
NM_175859.2 | NP_787055.1 | 4 | 787 | |||
XM_005274562.3 | XP_005274619.1 | 4 | 564 | |||
XM_006724503.3 | XP_006724566.1 | 5 | 739 | |||
XM_011545545.2 | XP_011543847.1 | 4 | 560 | |||
GenBank mRNA | AF226667.1 | - | 4 | 604 | ||
AK023549.1 | - | 4 | 659 | |||
AK024070.1 | - | 4 | 642 | |||
AK125332.1 | - | 4 | 1148 | |||
AK125348.1 | - | 4 | 778 | |||
BC006256.2 | - | 2 | 143 | |||
BC034986.1 | - | 4 | 766 | 5268973 |