Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EIF4ENIF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164501.1 | NP_001157973.1 | 5 | 710 | ||
NM_019843.3 | NP_062817.2 | 5 | 734 | |||
XM_005261686.2 | XP_005261743.1 | 5 | 734 | |||
XM_005261687.2 | XP_005261744.1 | 5 | 715 | |||
XM_005261688.2 | XP_005261745.1 | 5 | 585 | |||
XM_006724281.3 | XP_006724344.3 | 5 | 1011 | |||
XM_011530280.1 | XP_011528582.1 | 5 | 585 | |||
XM_011530281.2 | XP_011528583.1 | 5 | 806 | |||
XM_017028863.1 | XP_016884352.1 | 5 | 1011 | |||
XM_017028864.1 | XP_016884353.1 | 5 | 806 | |||
GenBank mRNA | AF240775.1 | - | 5 | 564 | ||
AK314636.1 | - | 5 | 735 | |||
BC012734.2 | - | 5 | 587 | |||
BC033028.1 | - | 5 | 684 | 4837225 | ||
CR456386.1 | - | 4 | 543 |