Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHST7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019886.3 | NP_063939.2 | 2 | 2132 | ||
GenBank mRNA | AB037187.1 | - | 2 | 1897 | ||
BC045537.1 | - | 2 | 2035 | 5261787 | ||
BP250418.1 | - | 1 | 240 | |||
DA277492.1 | - | 1 | 242 |