Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP26B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001277742.1 | NP_001264671.1 | 4 | 725 | ||
NM_019885.3 | NP_063938.1 | 5 | 950 | |||
XM_005264433.4 | XP_005264490.1 | 5 | 914 | |||
XM_011532988.1 | XP_011531290.1 | 4 | 511 | |||
GenBank mRNA | AF252297.1 | - | 5 | 922 | ||
AK294814.1 | - | 4 | 697 | |||
AK294933.1 | - | 4 | 920 | |||
AK295683.1 | - | 4 | 565 | |||
AK313433.1 | - | 5 | 925 | |||
BC069443.1 | - | 5 | 922 | 7262360 | ||
BC109205.2 | - | 5 | 922 | 40006245 | ||
CR749631.1 | - | 4 | 574 | |||
FJ467289.1 | - | 4 | 697 |