Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB8P7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC014971.1 | - | 3 | 1769 | ||
BC139719.1 | - | 4 | 1343 |
Gene Symbol: TUBB8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177987.2 | NP_817124.1 | 4 | 1152 | ||
XM_011519459.2 | XP_011517761.1 | 4 | 1192 | |||
XM_011519460.2 | XP_011517762.1 | 3 | 838 | |||
XM_017016192.1 | XP_016871681.1 | 2 | 1089 | |||
XM_017016193.1 | XP_016871682.1 | 2 | 909 | |||
GenBank mRNA | BC101270.2 | - | 4 | 1145 | ||
BC101271.1 | - | 4 | 1145 |
Gene Symbol: TUBBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027156.1 | 5 | 2308 |