Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083947.1 | NP_001077416.1 | 12 | 1546 | ||
NM_001173551.1 | NP_001167022.1 | 12 | 1555 | |||
NM_001173552.1 | NP_001167023.1 | 11 | 1441 | |||
NM_001290094.1 | NP_001277023.1 | 12 | 1565 | |||
NM_001290096.1 | NP_001277025.1 | 12 | 1476 | |||
NM_019894.3 | NP_063947.1 | 12 | 1561 | |||
NR_110734.1 | 11 | 1412 | ||||
XM_005271613.3 | XP_005271670.1 | 12 | 1554 | |||
XM_005271614.2 | XP_005271671.1 | 12 | 1519 | |||
XM_005271615.3 | XP_005271672.1 | 11 | 1436 | |||
XM_011542901.2 | XP_011541203.1 | 12 | 1541 | |||
XM_011542902.2 | XP_011541204.1 | 11 | 1442 | |||
GenBank mRNA | AF179224.1 | - | 12 | 1484 | ||
AF216312.1 | - | 12 | 1478 | |||
AK172766.1 | - | 12 | 1417 | |||
AK290361.1 | - | 11 | 1346 | |||
AK296855.1 | - | 12 | 1414 | |||
AK303173.1 | - | 11 | 1376 | |||
AK304113.1 | - | 12 | 1529 | |||
AK304215.1 | - | 12 | 1499 | |||
AK308434.1 | - | 12 | 1497 | |||
AY358530.1 | - | 12 | 1464 | |||
BC004855.1 | - | 11 | 1363 | 3835263 | ||
BC011703.2 | - | 12 | 1477 | 3610695 |