Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163812.1 | NP_001157284.1 | 1 | 1225 | ||
NM_021830.4 | NP_068602.2 | 1 | 1225 | |||
XM_017016437.1 | XP_016871926.1 | 1 | 724 | |||
GenBank mRNA | AF292004.1 | - | 1 | 735 | ||
AF292005.1 | - | 1 | 735 | |||
AK025485.1 | - | 1 | 48 | |||
AK308098.1 | - | 1 | 1225 | |||
BC013349.2 | - | 1 | 158 | |||
BC033762.1 | - | 1 | 218 | 3854007 | ||
BX640829.1 | - | 1 | 1207 |