Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C10orf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163812.1 | NP_001157284.1 | 5 | 2579 | ||
NM_001163813.1 | NP_001157285.1 | 5 | 695 | |||
NM_001163814.1 | NP_001157286.1 | 5 | 739 | |||
NM_021830.4 | NP_068602.2 | 5 | 2535 | |||
XM_011539975.2 | XP_011538277.1 | 5 | 642 | |||
XM_017016437.1 | XP_016871926.1 | 5 | 1972 | |||
GenBank mRNA | AF292004.1 | - | 5 | 2045 | ||
AF292005.1 | - | 5 | 2089 | |||
AK022959.1 | - | 1 | 13 | |||
AK025485.1 | - | 5 | 1358 | |||
AK297068.1 | - | 5 | 688 | |||
BC013349.2 | - | 5 | 1512 | |||
BC033762.1 | - | 5 | 1528 | 3854007 | ||
BQ643363.1 | - | 5 | 681 | |||
BX640829.1 | - | 5 | 2637 |