Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLE4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019896.2 | NP_063949.2 | 4 | 472 | ||
GenBank mRNA | BC031331.1 | - | 4 | 467 | 5443810 | |
BU597500.1 | - | 479 | ||||
CA454961.1 | - | 4 | 414 |