Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACKR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001296.4 | NP_001287.2 | 3 | 390 | ||
GenBank mRNA | AK313561.1 | - | 3 | 387 | ||
BC008816.2 | - | 3 | 378 | 3959017 | ||
BC011631.2 | - | 3 | 378 | 3958671 | ||
BC020558.1 | - | 2 | 309 | 3451472 | ||
BC101629.1 | - | 1 | 242 | 8069135 | ||
BC112045.1 | - | 1 | 242 | 8327513 | ||
BE901130.1 | - | 3 | 359 | |||
BT006800.1 | - | 1 | 211 | |||
U94888.1 | - | 1 | 226 | |||
Y12815.1 | - | 1 | 223 |