Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206646.1 | NP_001193575.1 | 3 | 348 | ||
NM_001206647.1 | NP_001193576.1 | 4 | 429 | |||
NM_001206648.1 | NP_001193577.1 | 3 | 348 | |||
NM_020642.3 | NP_065693.2 | 4 | 429 | |||
NR_045417.1 | 3 | 611 | ||||
NR_045418.1 | 2 | 530 | ||||
XM_017018011.1 | XP_016873500.1 | 4 | 449 | |||
XM_017018012.1 | XP_016873501.1 | 3 | 376 | |||
GenBank mRNA | AF493783.1 | - | 4 | 429 | ||
AF493784.1 | - | 4 | 429 | |||
AF493785.1 | - | 3 | 348 | |||
AF512007.1 | - | 3 | 353 | |||
AK075308.1 | - | 548 | ||||
AK294506.1 | - | 2 | 521 | |||
AK294549.1 | - | 3 | 590 | |||
AW340643.1 | - | 2 | 203 | |||
BC030996.2 | - | 3 | 359 | 4609949 | ||
BM706322.1 | - | 2 | 384 |