Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC2A4RG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020062.3 | NP_064446.2 | 8 | 1417 | ||
GenBank mRNA | AB044777.1 | - | 7 | 1309 | ||
AB044786.1 | - | 6 | 978 | |||
AB052777.1 | - | 8 | 1461 | |||
AF249267.3 | - | 8 | 1362 | |||
AK298304.1 | - | 6 | 1340 | |||
BC001402.2 | - | 7 | 1188 | |||
BC017446.1 | - | 5 | 1011 | |||
BC028349.2 | - | 3 | 612 | |||
BC052306.1 | - | 8 | 1462 | 6576325 |