Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BARHL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020064.3 | NP_064448.1 | 1 | 481 | ||
GenBank mRNA | AF321618.1 | - | 1 | 289 | ||
AF325688.1 | - | 1 | 304 | |||
AK054797.1 | - | 1 | 481 | |||
BC136956.1 | - | 1 | 428 | 9020948 | ||
BC136969.1 | - | 1 | 428 | 9020961 | ||
BC144433.1 | - | 2 | 341 | 9052955 |