Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FMN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305424.1 | NP_001292353.1 | 6 | 4145 | ||
NM_020066.4 | NP_064450.3 | 5 | 4133 | |||
XM_011544237.2 | XP_011542539.1 | 6 | 4143 | |||
XM_017001837.1 | XP_016857326.1 | 6 | 4143 | |||
XM_017001838.1 | XP_016857327.1 | 6 | 4143 | |||
XM_017001839.1 | XP_016857328.1 | 6 | 2985 | |||
XM_017001840.1 | XP_016857329.1 | 5 | 2385 | |||
XM_017001841.1 | XP_016857330.1 | 5 | 2843 | |||
GenBank mRNA | AB209153.1 | - | 6 | 2738 | ||
DN990332.1 | - | 1 | 137 |