Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGGT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020120.3 | NP_064505.1 | 30 | 3532 | ||
NR_027671.2 | 30 | 3685 | ||||
XM_006712634.3 | XP_006712697.1 | 30 | 3506 | |||
XM_006712635.3 | XP_006712698.1 | 29 | 3476 | |||
XM_006712636.3 | XP_006712699.1 | 30 | 3495 | |||
XM_017004507.1 | XP_016859996.1 | 31 | 3591 | |||
XM_017004508.1 | XP_016859997.1 | 31 | 3546 | |||
XM_017004509.1 | XP_016859998.1 | 16 | 1837 | |||
XM_017004510.1 | XP_016859999.1 | 12 | 2216 | |||
GenBank mRNA | AF227905.1 | - | 30 | 3392 | ||
AK023671.1 | - | 11 | 1228 | |||
AK074251.1 | - | 14 | 1556 | |||
AK074376.1 | - | 4 | 527 | |||
AK293066.1 | - | 30 | 3685 | |||
BC041098.1 | - | 30 | 3682 | 5492334 |