Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCMF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020122.4 | NP_064507.3 | 2 | 421 | ||
XM_006712052.3 | XP_006712115.1 | 2 | 453 | |||
XM_011532990.1 | XP_011531292.1 | 2 | 343 | |||
GenBank mRNA | AB083199.1 | - | 2 | 130 | ||
AF155652.1 | - | 2 | 130 | |||
AK023403.1 | - | 2 | 100 | |||
AK314761.1 | - | 2 | 196 | |||
BC000178.2 | - | 2 | 220 | 3048124 | ||
BU844982.1 | - | 2 | 405 | |||
CT004343.1 | - | 2 | 421 |