Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBQLN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304342.1 | NP_001291271.1 | 10 | 1704 | ||
NM_020131.4 | NP_064516.2 | 10 | 1764 | |||
GenBank mRNA | AB209566.1 | - | 10 | 1637 | ||
AF113544.1 | - | 7 | 1170 | |||
AF188240.1 | - | 10 | 1918 | |||
AK098368.1 | - | 5 | 609 | |||
AK302895.1 | - | 10 | 1593 | |||
AK314413.1 | - | 10 | 1652 | |||
BC006410.1 | - | 1 | 39 | |||
BC018403.2 | - | 10 | 1652 | 4214386 | ||
BC063841.1 | - | 10 | 1664 | 6183942 |