Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WRNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020135.2 | NP_064520.2 | 2 | 1184 | ||
NM_130395.2 | NP_569079.1 | 2 | 1184 | |||
XM_005249232.3 | XP_005249289.1 | 2 | 1275 | |||
XM_017011060.1 | XP_016866549.1 | 2 | 1275 | |||
GenBank mRNA | AB056152.1 | - | 2 | 1185 | ||
AB209723.1 | - | 2 | 937 | |||
AF218313.1 | - | 2 | 571 | |||
AK026179.1 | - | 1 | 89 | |||
AK094823.1 | - | 2 | 1144 | |||
AK223593.1 | - | 2 | 1118 | |||
AK315047.1 | - | 2 | 1276 | |||
AK315471.1 | - | 2 | 1153 | |||
BC018923.2 | - | 2 | 1145 | 4109718 |