Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM167B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001322248.1 | NP_001309177.1 | 2 | 185 | ||
NM_020141.3 | NP_064526.1 | 2 | 145 | |||
NR_136242.1 | 3 | 274 | ||||
GenBank mRNA | AF164793.1 | - | 2 | 166 | ||
AK075191.1 | - | 2 | 167 | |||
AK291293.1 | - | 2 | 135 | |||
BC008739.2 | - | 2 | 145 | 3346340 | ||
BC065186.1 | - | 2 | 145 | 4642864 | ||
BC066918.1 | - | 2 | 174 | 4826492 | ||
BC067116.1 | - | 2 | 132 | 5532892 | ||
BC146873.1 | - | 2 | 1123 | 9056801 | ||
BC146879.1 | - | 2 | 1123 | 9056807 | ||
BC171849.1 | - | 2 | 1123 | 9054503 | ||
BP240791.1 | - | 2 | 153 | |||
BX331284.2 | - | 3 | 235 |