Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NDUFA4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020142.3 | NP_064527.1 | 4 | 634 | ||
XM_005269033.2 | XP_005269090.1 | 6 | 607 | |||
XM_011538573.2 | XP_011536875.1 | 5 | 1249 | |||
GenBank mRNA | AF164796.1 | - | 4 | 379 | ||
AK094275.1 | - | 8 | 1464 | |||
AK129629.1 | - | 4 | 651 | |||
BC011910.2 | - | 4 | 634 | 4138587 |