Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286555.1 | NP_001273484.1 | 6 | 792 | ||
NM_020185.4 | NP_064570.1 | 6 | 792 | |||
NR_104473.1 | 5 | 742 | ||||
NR_104474.1 | 5 | 709 | ||||
NR_104475.1 | 3 | 625 | ||||
XM_011514757.2 | XP_011513059.1 | 3 | 254 | |||
XM_017011062.1 | XP_016866551.1 | 3 | 226 | |||
GenBank mRNA | AB208997.1 | - | 3 | 145 | ||
AF165519.1 | - | 6 | 333 | |||
AF424702.1 | - | 6 | 392 | |||
AK000383.1 | - | 2 | 118 | |||
AK074410.1 | - | 2 | 118 | |||
AK128760.1 | - | 3 | 1113 | |||
AK296402.1 | - | 6 | 362 | |||
AK310653.1 | - | 5 | 321 | |||
AY249859.1 | - | 6 | 796 | |||
BC016844.1 | - | 1 | 51 | |||
BC022847.2 | - | 6 | 748 | 3942055 | ||
BX362191.2 | - | 3 | 406 | |||
BX648953.1 | - | 3 | 4791 | |||
DA950312.1 | - | 5 | 325 | |||
DB177174.1 | - | 5 | 246 |