Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193557.1 | NP_001180486.1 | 2 | 125 | ||
NM_020189.5 | NP_064574.1 | 2 | 197 | |||
NR_036471.1 | 2 | 197 | ||||
NR_036472.1 | 2 | 197 | ||||
GenBank mRNA | AF173296.1 | - | 2 | 267 | ||
AF201940.1 | - | 2 | 212 | |||
AK000487.1 | - | 2 | 101 | |||
AK095651.1 | - | 2 | 196 | |||
AK290752.1 | - | 2 | 141 | |||
AK316560.1 | - | 2 | 192 | |||
BC007870.2 | - | 2 | 113 | 4303290 | ||
CR457183.1 | - | 1 | 51 | |||
DA548665.1 | - | 2 | 71 | |||
DA762166.1 | - | 2 | 179 | |||
DB168034.1 | - | 2 | 197 |