Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OLFML3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286352.1 | NP_001273281.1 | 3 | 603 | ||
NM_001286353.1 | NP_001273282.1 | 2 | 580 | |||
NM_020190.3 | NP_064575.1 | 2 | 451 | |||
XM_017001848.1 | XP_016857337.1 | 2 | 613 | |||
GenBank mRNA | AF201945.1 | - | 2 | 385 | ||
AK075353.1 | - | 2 | 353 | |||
AK075369.1 | - | 2 | 399 | |||
AK075479.1 | - | 2 | 364 | |||
AK075544.1 | - | 2 | 351 | |||
AK092766.1 | - | 3 | 553 | |||
AK222471.1 | - | 2 | 368 | |||
AK223221.1 | - | 2 | 381 | |||
AK297902.1 | - | 2 | 287 | |||
AY358954.1 | - | 2 | 434 | |||
AY464015.1 | - | 2 | 416 | |||
BC009920.2 | - | 2 | 376 | 3010136 | ||
BP201458.1 | - | 2 | 497 | |||
KC237062.1 | - | 2 | 325 |