Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EMSY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300942.1 | NP_001287871.1 | 16 | 2485 | ||
NM_001300943.1 | NP_001287872.1 | 15 | 2443 | |||
NM_001300944.1 | NP_001287873.1 | 16 | 2485 | |||
NM_020193.4 | NP_064578.2 | 15 | 2440 | |||
XM_005274112.4 | XP_005274169.1 | 15 | 2429 | |||
XM_006718638.3 | XP_006718701.1 | 15 | 2453 | |||
XM_011545173.2 | XP_011543475.1 | 16 | 2582 | |||
XM_011545174.2 | XP_011543476.1 | 16 | 2579 | |||
XM_011545175.2 | XP_011543477.1 | 15 | 2453 | |||
XM_011545177.2 | XP_011543479.1 | 15 | 2429 | |||
XM_011545179.2 | XP_011543481.1 | 14 | 2300 | |||
XM_017018021.1 | XP_016873510.1 | 16 | 2582 | |||
XM_017018022.1 | XP_016873511.1 | 16 | 2428 | |||
GenBank mRNA | AJ430203.1 | - | 14 | 2297 | ||
AK125114.1 | - | 14 | 2154 | |||
AK126030.1 | - | 3 | 459 | |||
AK304106.1 | - | 3 | 350 | |||
BC117265.1 | - | 14 | 2339 | 40125816 | ||
BC143374.1 | - | 15 | 2381 | 9051879 | ||
BC143376.1 | - | 15 | 2381 | 9051881 | ||
BX647398.1 | - | 4 | 535 |