Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XAB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020196.2 | NP_064581.2 | 13 | 1719 | ||
GenBank mRNA | AB026111.1 | - | 13 | 1708 | ||
AB033003.1 | - | 11 | 1395 | |||
AF226051.1 | - | 13 | 1722 | |||
AF258567.1 | - | 13 | 1714 | |||
AF272147.1 | - | 13 | 1729 | |||
AK025858.1 | - | 8 | 1002 | |||
AK074035.1 | - | 8 | 3323 | |||
BC007208.1 | - | 13 | 1701 | 2823235 | ||
BC008778.1 | - | 8 | 1034 | |||
CR749864.1 | - | 10 | 1248 |