Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRTFDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020200.6 | NP_064585.1 | 8 | 666 | ||
GenBank mRNA | AB209792.1 | - | 8 | 616 | ||
AF226056.1 | - | 8 | 614 | |||
AK021950.1 | - | 8 | 626 | |||
AK222674.1 | - | 8 | 625 | |||
BC008662.2 | - | 8 | 628 | 3871911 |