Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NIT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020202.4 | NP_064587.1 | 4 | 389 | ||
GenBank mRNA | AF260334.1 | - | 4 | 388 | ||
AF284574.1 | - | 4 | 336 | |||
AK225979.1 | - | 4 | 365 | |||
AK294010.1 | - | 4 | 350 | |||
AK295835.1 | - | 4 | 350 | |||
AK313704.1 | - | 4 | 331 | |||
BC020620.1 | - | 4 | 346 | 4775062 | ||
BC107890.1 | - | 4 | 377 | 6648910 | ||
DB466042.1 | - | 4 | 389 | |||
FJ224326.1 | - | 4 | 336 |