Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284395.1 | NP_001271324.1 | 16 | 1819 | ||
NM_020212.1 | NP_064597.1 | 16 | 1903 | |||
XM_006720614.1 | XP_006720677.1 | 15 | 1825 | |||
XM_006720615.1 | XP_006720678.1 | 14 | 1738 | |||
XM_011521794.2 | XP_011520096.1 | 15 | 2118 | |||
XM_011521795.1 | XP_011520097.1 | 15 | 1849 | |||
XM_011521798.2 | XP_011520100.1 | 14 | 2109 | |||
XM_011521800.1 | XP_011520102.1 | 13 | 1279 | |||
XM_017022415.1 | XP_016877904.1 | 14 | 1742 | |||
XM_017022416.1 | XP_016877905.1 | 12 | 1209 | |||
GenBank mRNA | AK097525.1 | - | 16 | 1787 | ||
AK302032.1 | - | 10 | 1308 | |||
AL390084.1 | - | 8 | 884 | |||
AL390085.1 | - | 8 | 884 | |||
BC064626.1 | - | 16 | 1903 | 5742386 |