Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POGLUT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152305.2 | NP_689518.1 | 3 | 352 | ||
NR_024265.1 | 3 | 352 | ||||
XM_006713705.3 | XP_006713768.1 | 2 | 163 | |||
XM_017006878.1 | XP_016862367.1 | 2 | 183 | |||
XM_017006879.1 | XP_016862368.1 | 3 | 298 | |||
XM_017006880.1 | XP_016862369.1 | 3 | 298 | |||
GenBank mRNA | AF168711.1 | - | 3 | 303 | ||
AK075444.1 | - | 3 | 288 | |||
AK222937.1 | - | 3 | 312 | |||
AK295984.1 | - | 3 | 327 | |||
AK301002.1 | - | 3 | 327 | |||
AK315367.1 | - | 3 | 327 | |||
AY298903.1 | - | 3 | 338 | |||
AY358581.1 | - | 3 | 295 | |||
BC030614.2 | - | 3 | 338 | 4796951 | ||
BC048810.1 | - | 3 | 296 | 5243487 | ||
BP347179.1 | - | 3 | 352 | |||
CD517010.1 | - | 3 | 268 |