Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020244.2 | NP_064629.2 | 5 | 922 | ||
XM_011538575.1 | XP_011536877.1 | 5 | 619 | |||
GenBank mRNA | AF047431.1 | - | 5 | 493 | ||
AF111803.1 | - | 4 | 357 | |||
AF195623.1 | - | 5 | 869 | |||
AF195624.1 | - | 6 | 896 | |||
AK075211.1 | - | 5 | 889 | |||
AK093067.1 | - | 6 | 782 | |||
AK226127.1 | - | 6 | 623 | |||
AY280609.1 | - | 3 | 172 | |||
AY280610.1 | - | 3 | 180 | |||
AY294627.1 | - | 3 | 180 | |||
BC020819.1 | - | 5 | 876 | 4696613 | ||
BC050429.1 | - | 5 | 915 | 6473945 |