Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC12A9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001267812.1 | NP_001254741.1 | 8 | 1202 | ||
NM_001267814.1 | NP_001254743.1 | 6 | 939 | |||
NM_020246.3 | NP_064631.2 | 8 | 1202 | |||
XM_005250502.2 | XP_005250559.1 | 6 | 938 | |||
XM_005250504.4 | XP_005250561.1 | 2 | 173 | |||
XM_006716054.2 | XP_006716117.1 | 6 | 920 | |||
XM_006716055.2 | XP_006716118.1 | 6 | 775 | |||
XM_011516414.1 | XP_011514716.1 | 5 | 608 | |||
GenBank mRNA | AB033284.1 | - | 8 | 1129 | ||
AF284422.1 | - | 8 | 1169 | |||
AK024420.1 | - | 4 | 1350 | |||
AK024421.1 | - | 4 | 2019 | |||
AK024466.1 | - | 1 | 1214 | |||
AK024494.1 | - | 3 | 2327 | |||
AK090458.1 | - | 4 | 1328 | |||
AK301411.1 | - | 8 | 1202 | |||
BC000154.2 | - | 6 | 921 | 2900382 | ||
CA488831.1 | - | 80 |