Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: YAE1D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020192.4 | NP_064577.1 | 3 | 471 | ||
GenBank mRNA | AF226046.1 | - | 3 | 437 | ||
AK314937.1 | - | 3 | 443 | |||
BC022043.1 | - | 3 | 441 | 4734433 | ||
BM767075.1 | - | 3 | 471 | |||
BX648555.1 | - | 1 | 3024 | |||
CR457198.1 | - | 3 | 428 |