Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CABP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300895.1 | NP_001287824.1 | 4 | 730 | ||
NM_001300896.1 | NP_001287825.1 | 5 | 614 | |||
NM_145200.3 | NP_660201.1 | 4 | 662 | |||
XM_005274114.3 | XP_005274171.2 | 5 | 642 | |||
XM_011545181.2 | XP_011543483.1 | 5 | 667 | |||
XM_011545182.2 | XP_011543484.1 | 4 | 523 | |||
XM_011545183.2 | XP_011543485.1 | 5 | 466 | |||
XM_017018025.1 | XP_016873514.1 | 5 | 952 | |||
GenBank mRNA | AY039217.1 | - | 4 | 662 | ||
BC033167.1 | - | 5 | 614 | 4549347 |