Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033133.4 | NP_149124.3 | 2 | 515 | ||
XM_006721701.3 | XP_006721764.1 | 2 | 539 | |||
XM_011524340.2 | XP_011522642.1 | 2 | 500 | |||
GenBank mRNA | AK026876.1 | - | 2 | 472 | ||
AK097833.1 | - | 1 | 470 | |||
AK098384.1 | - | 2 | 461 | |||
BC001362.2 | - | 2 | 452 | 3051112 | ||
BC006392.2 | - | 2 | 474 | 4126696 | ||
BC011046.1 | - | 2 | 436 | 4179083 | ||
BC028040.1 | - | 2 | 476 | 5248370 | ||
CB988051.1 | - | 2 | 542 | |||
M19650.1 | - | 1 | 324 |