Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CIAPIN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308347.1 | NP_001295276.1 | 5 | 716 | ||
NM_001308358.1 | NP_001295287.1 | 5 | 755 | |||
NM_020313.3 | NP_064709.2 | 5 | 755 | |||
XM_005256063.4 | XP_005256120.1 | 5 | 607 | |||
GenBank mRNA | AF116609.1 | - | 3 | 366 | ||
AF248964.1 | - | 5 | 659 | |||
AK074263.1 | - | 5 | 596 | |||
AK292281.1 | - | 5 | 559 | |||
AK295024.1 | - | 4 | 988 | |||
AL136613.1 | - | 5 | 577 | |||
BC002568.2 | - | 5 | 597 | 3139226 | ||
BC024196.2 | - | 5 | 636 | 4816594 | ||
BC067303.1 | - | 5 | 755 | 30389410 | ||
BC071740.1 | - | 5 | 630 | 5563094 | ||
BG746807.1 | - | 1 | 601 | |||
CR533545.1 | - | 4 | 474 |