Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RSRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321772.1 | NP_001308701.1 | 3, 4 | 1959 | ||
NM_020317.4 | NP_064713.3 | 3, 4 | 955 | |||
XM_011541797.1 | XP_011540099.1 | 3, 4 | 920 | |||
GenBank mRNA | BC001892.1 | - | 3, 4 | 879 | 3534629 | |
BC065040.1 | - | 3, 4 | 868 | 6147639 |