Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001160226.1 | NP_001153698.1 | 3 | 1308 | ||
NM_001160258.1 | NP_001153730.1 | 4 | 1346 | |||
NM_001160259.1 | NP_001153731.1 | 2 | 1221 | |||
NM_016083.4 | NP_057167.2 | 2 | 1165 | |||
NM_033181.3 | NP_149421.2 | 2 | 820 | |||
XM_006715330.3 | XP_006715393.1 | 2 | 1112 | |||
XM_011535424.1 | XP_011533726.1 | 3 | 1652 | |||
XM_011535425.1 | XP_011533727.1 | 4 | 1690 | |||
XM_011535426.1 | XP_011533728.1 | 5 | 1848 | |||
XM_011535428.1 | XP_011533730.1 | 3 | 1499 | |||
XM_017010238.1 | XP_016865727.1 | 4 | 4476 | |||
XM_017010239.1 | XP_016865728.1 | 3 | 2799 | |||
XM_017010240.1 | XP_016865729.1 | 2 | 1159 | |||
GenBank mRNA | AF107262.1 | - | 1 | 865 | ||
AK313908.1 | - | 2 | 986 | |||
AY766182.1 | - | 2 | 757 | |||
AY995204.1 | - | 1 | 856 | |||
BC074811.2 | - | 1 | 894 | 30915316 | ||
BC074812.2 | - | 1 | 894 | 30915532 | ||
BC095513.1 | - | 1 | 853 | |||
BC100968.1 | - | 1 | 856 | 40004970 | ||
BC100969.1 | - | 1 | 856 | 40004975 | ||
BC100970.1 | - | 1 | 856 | 40004978 | ||
BC100971.1 | - | 1 | 856 | 40004979 | ||
DQ067455.1 | - | 1 | 856 | |||
KC292264.1 | - | 2 | 820 | |||
X54937.1 | - | 2 | 1004 | |||
X81120.1 | - | 1 | 947 | |||
X81121.1 | - | 2 | 689 |
Set Membership: |
G-protein coupled receptor |